Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01214968_m1
          See other TTC9C GE Assays ›
          Gene Symbol:
          Ttc9c
          Gene Name:
          tetratricopeptide repeat domain 9C
          Gene Aliases:
          2210019E14Rik, 6330408J23Rik
          Chromosome Location:
          Chr.19: 8809066 - 8819780 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01214968_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc9c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027412.3 NP_081688.2 3 - 4 708 68
          GenBank mRNA AK008753.1 - 2 - 3 1039 68
          AK018145.1 - 3 - 4 748 68
          AK169288.1 - 3 - 4 709 68
          AK169296.1 - 3 - 4 709 68
          BC035304.1 - 2 - 3 619 68
          BC043061.1 - 2 - 3 1020 5704924 68
          BC058967.1 - 2 - 3 1020 5704924 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001071.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ttc9c

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 BC035304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK169288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK169296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK039321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 NM_027412.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 BC035304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK169288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK169296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 AK039321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P531662 NM_027412.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC063773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC035304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC043061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC058967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 AK169288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC063773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC035304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC043061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 BC058967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P178872 AK169288.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90857 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 BC035304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK169288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK169296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK039321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 NM_027412.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 BC035304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK169288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK169296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 AK039321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P531662 NM_027412.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC063773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC035304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC043061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC058967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 AK169288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC063773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC035304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC043061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 BC058967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P178872 AK169288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 BC035304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK169288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK169296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK039321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 NM_027412.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 BC035304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK169288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK169296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 AK039321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531662 NM_027412.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC063773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC035304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC043061.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC058967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 AK169288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC063773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC035304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC043061.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 BC058967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178872 AK169288.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465816 - -
          Affymetrix HT MG-430 PM Array Plate 1443948_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434001_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432245_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1430569_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465812 - -
          Mouse Genome 430 2.0 Array 1432245_PM_s_at - -
          Mouse Genome 430 2.0 Array 1430569_at - -
          Mouse Genome 430 2.0 Array 1443948_PM_at - -
          Mouse Genome 430 2.0 Array 1434001_PM_at - -
          Mouse Genome 430 2.0 Array 1432245_s_at - -
          Mouse Genome 430 2.0 Array 1434001_at - -
          Mouse Genome 430 2.0 Array 1430569_PM_at - -
          Mouse Genome 430 2.0 Array 1443948_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70387_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70387_at - -
          Affymetrix Mouse Clariom S Array PSR1900010040.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010040.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010040.mm.1 TC1900001071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010041.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010041.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010041.mm.1 TC1900001071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010043.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010043.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010043.mm.1 TC1900001071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010045.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010045.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010045.mm.1 TC1900001071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010038.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010038.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010038.mm.1 TC1900001071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010039.mm.1 NM_027412.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010039.mm.1 NM_027412.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010039.mm.1 TC1900001071.mm.1 MTA TC
          Mouse Expression Set 430 1430569_PM_at - -
          Mouse Expression Set 430 1443948_at - -
          Mouse Expression Set 430 1434001_PM_at - -
          Mouse Expression Set 430 1434001_at - -
          Mouse Expression Set 430 1443948_PM_at - -
          Mouse Expression Set 430 1430569_at - -
          Murine Genome U74v2 Set 168388_at - -
          Murine Genome U74v2 Set 168388_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434001_at - -
          Affymetrix HT Mouse Genome 430B Array 1432245_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430569_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443948_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434001_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443948_at - -
          Affymetrix HT Mouse Genome 430B Array 1432245_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 648619 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233738 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223685 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline