Search
Search

Ppdpf| Gene Symbol: Ppdpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025598.2 | NP_079874.1 | 2 - 3 | 297 | 83 | |
| GenBank mRNA | AK003282.1 | - | 2 - 3 | 298 | 83 | |
| AK012037.1 | - | 2 - 3 | 181 | 83 | ||
| AK012327.1 | - | 2 - 3 | 299 | 83 | ||
| AK154758.1 | - | 2 - 3 | 338 | 83 | ||
| BC058516.1 | - | 2 - 3 | 335 | 6824261 | 83 | |
| BC091753.1 | - | 2 - 3 | 104 | 4218108 | 83 | |
| BC100439.1 | - | 2 - 3 | 189 | 6531448 | 83 | |
Set Membership: |
|