Search
Search

Mrgbp| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028479.1 | NP_082755.1 | 3 - 4 | 699 | 74 | |
| GenBank mRNA | AK005551.1 | - | 2 - 3 | 339 | 74 | |
| AK052575.1 | - | 2 - 3 | 354 | 74 | ||
| AK217601.1 | - | 1 - 2 | 116 | 74 | ||
| BC054431.1 | - | 2 - 3 | 286 | 74 | ||
| BC116732.1 | - | 3 - 4 | 699 | 40126051 | 74 | |
| BC116734.1 | - | 3 - 4 | 699 | 40126053 | 74 | |
Set Membership: |
|