Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215175_m1
          See other GID8 GE Assays ›
          Gene Symbol:
          Gid8
          Gene Name:
          GID complex subunit 8 homolog (S. cerevisiae)
          Gene Aliases:
          2310003C23Rik, 4833420G11Rik, AI451474, Twa1
          Chromosome Location:
          Chr.2: 180710226 - 180721599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215175_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gid8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289651.1 NP_001276580.1 4 - 5 685 88
          NM_001289652.1 NP_001276581.1 4 - 5 651 88
          NM_029607.2 NP_083883.1 3 - 4 1171 88
          GenBank mRNA AK009106.1 - 4 - 5 611 88
          BC052539.1 - 4 - 5 683 88
          BC059022.1 - 3 - 4 1171 6415235 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002801.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gid8

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040628 BC052539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040628 BC059022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040628 NM_029607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040628 AK009106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040628 AK211027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125335 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040628 BC052539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040628 BC059022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040628 NM_029607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040628 AK009106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040628 AK211027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21905722 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125335 - -
          Whole Mouse Genome G4122A A_51_P133445 BC052539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133445 BC059022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133445 NM_029607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133445 AK009106.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02226415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02226413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21905818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02226413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01480363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02241318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01685874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01336690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02143203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01336687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01685873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21905726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10479460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479462 - -
          Affymetrix HT MG-430 PM Array Plate 1428824_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10479458 - -
          Mouse Genome 430 2.0 Array 1428824_PM_at - -
          Mouse Genome 430 2.0 Array 1428824_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76425_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76425_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200022945.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022945.mm.1 TC0200002801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022941.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022941.mm.1 TC0200002801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022944.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022944.mm.1 TC0200002801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022942.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022942.mm.1 TC0200002801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022946.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022946.mm.1 TC0200002801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022943.mm.1 NM_029607.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022943.mm.1 TC0200002801.mm.1 MTA TC
          Mouse Expression Set 430 1428824_at - -
          Mouse Expression Set 430 1428824_PM_at - -
          Murine Genome U74v2 Set 165974_r_at - -
          Murine Genome U74v2 Set 165974_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1428824_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428824_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 380184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2730729 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244393 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2730729 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244393 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244393 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline