Search
Search

Ddx58| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 14 - 15 | 2141 | 65 | |
| GenBank mRNA | AK049305.1 | - | 5 - 6 | 732 | 65 | |
| AK052595.1 | - | 10 - 11 | 1395 | 65 | ||
| AK128929.1 | - | 14 - 15 | 2076 | 65 | ||
| AK155930.1 | - | 14 - 15 | 2077 | 65 | ||
| AY553221.1 | - | 14 - 15 | 2017 | 65 | ||
| BC027369.1 | - | 14 - 15 | 2120 | 65 | ||
| BC106095.1 | - | 14 - 15 | 2061 | 65 | ||
| BC127163.1 | - | 14 - 15 | 2007 | 65 | ||
| BC152540.1 | - | 14 - 15 | 2009 | 65 | ||
Set Membership: |
|