Search
Search

Cdc16| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027276.2 | NP_081552.2 | 6 - 7 | 650 | 66 | |
| GenBank mRNA | AK147938.1 | - | 6 - 7 | 665 | 66 | |
| AK167965.1 | - | 6 - 7 | 665 | 66 | ||
| AK168978.1 | - | 6 - 7 | 637 | 66 | ||
| AK184740.1 | - | 2 - 3 | 243 | 66 | ||
| BC026606.1 | - | 6 - 7 | 650 | 4976424 | 66 | |
Set Membership: |
|