Search
Search

Fam193a| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243123.1 | NP_001230052.1 | 6 - 7 | 1427 | 55 | |
| GenBank mRNA | AK144988.1 | - | 5 - 6 | 944 | 55 | |
| BC037112.1 | - | 3 - 4 | 564 | 55 | ||
| BC099925.1 | - | 3 - 4 | 464 | 30653351 | 55 | |
Set Membership: |
|