Search
Search

Calb1| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009788.4 | NP_033918.1 | 6 - 7 | 584 | 95 | |
| GenBank mRNA | AK002635.1 | - | 6 - 7 | 586 | 95 | |
| AK005081.1 | - | 6 - 7 | 583 | 95 | ||
| AK005243.1 | - | 6 - 7 | 584 | 95 | ||
| AK028050.1 | - | 6 - 7 | 585 | 95 | ||
| AK028327.1 | - | 6 - 7 | 95 | |||
| AK048517.1 | - | 6 - 7 | 585 | 95 | ||
| AK077499.1 | - | 6 - 7 | 583 | 95 | ||
| AK078358.1 | - | 6 - 7 | 582 | 95 | ||
| BC016421.1 | - | 6 - 7 | 515 | 4527000 | 95 | |
| M21531.1 | - | 6 - 7 | 564 | 95 | ||
| M23663.1 | - | 3 - 4 | 146 | 95 | ||
Set Membership: |
|