Search
Search

Defb12| Gene Symbol: Defb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152802.3 | NP_690015.1 | 1 - 2 | 106 | 102 | |
| GenBank mRNA | AB089182.1 | - | 1 - 2 | 106 | 102 | |
| AK020311.1 | - | 1 - 2 | 104 | 102 | ||
Set Membership: |
|