Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218105_m1
          See other SMIM4 GE Assays ›
          Gene Symbol:
          Smim4
          Gene Name:
          small integral membrane protein 4
          Gene Aliases:
          2010107H07Rik, Gm41122
          Chromosome Location:
          Chr.14: 31124505 - 31128930 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218105_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007443.1 - 1 - 2 295 100
          BX630839.1 - 1 - 2 100
          CX238554.1 - 277 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563340 NR_024069.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563340 BX630839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563340 NR_024069.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P563340 BX630839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001903 NR_024069.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001903 AK019298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001903 BX630839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88771 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105636 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126982 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02824822 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08071864 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126982 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08071788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08071812 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126982 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418503 - -
          Affymetrix HT MG-430 PM Array Plate 1456204_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418500 - -
          Mouse Genome 430 2.0 Array 1456204_at - -
          Mouse Genome 430 2.0 Array 1456204_PM_at - -
          Mouse Expression Set 430 1456204_at - -
          Mouse Expression Set 430 1456204_PM_at - -
          Murine Genome U74v2 Set 93959_at - -
          Murine Genome U74v2 Set 93959_PM_at - -
          Murine Genome U74v2 Set 166440_i_at - -
          Murine Genome U74v2 Set 166440_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1456204_at - -
          Affymetrix HT Mouse Genome 430B Array 1456204_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678434 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669608 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669608 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline