Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218400_g1
          See other GALT GE Assays ›
          Gene Symbol:
          Galt
          Gene Name:
          galactose-1-phosphate uridyl transferase
          Gene Aliases:
          AW553376
          Chromosome Location:
          Chr.4: 41755228 - 41759224 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218400_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK088379.1 - 9 - 10 950 70
          AK134525.1 - 6 - 7 604 70
          AK163544.1 - 6 - 7 636 70
          AV027352.2 - 3 - 4 388 70
          BC010985.1 - 9 - 10 955 4038194 70
          M96265.1 - 979 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864174 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864173 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864172 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149269 NM_016658.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149269 AK134525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066927 AK088379.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73484 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118767 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149269 NM_016658.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149269 AK134525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117777 M96265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117777 NM_016658.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117777 AK134525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117777 AK163544.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P117777 AV027352.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22864183 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118767 - -
          Whole Mouse Genome G4122A A_51_P117777 M96265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117777 NM_016658.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117777 AK134525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117777 AK163544.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P117777 AV027352.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118581 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137552 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118767 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492101 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03736449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492089 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03736448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492091 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03492107 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864173 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864172 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22864174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22864189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04022732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03736448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03930105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04022735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04022733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03492103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22864183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04022730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504102 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504105 - -
          Affymetrix HT MG-430 PM Array Plate 1450187_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504094 - -
          Mouse Genome 430 2.0 Array 1450187_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450187_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14430_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14430_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400002416.mm.1 NM_016658.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400002416.mm.1 TC0400000382.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400002413.mm.1 NM_016658.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400002413.mm.1 TC0400000382.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400002414.mm.1 NM_016658.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400002414.mm.1 TC0400000382.mm.1 MTA TC
          Mouse Expression Set 430 1450187_PM_a_at - -
          Mouse Expression Set 430 1450187_a_at - -
          Murine Genome U74v2 Set 104616_PM_g_at - -
          Murine Genome U74v2 Set 104615_PM_at - -
          Murine Genome U74v2 Set 104615_at - -
          Murine Genome U74v2 Set 104616_g_at - -
          Mouse Genome 430A 2.0 Array 1450187_a_at - -
          Mouse Genome 430A 2.0 Array 1450187_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450187_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450187_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550075 - -
          MouseWG-6 v2 Expression BeadChip 3710162 - -
          Illumina mouse-6 v1.0 expression beadchip 5550075 - -
          Illumina mouse-6 v1.0 expression beadchip 3710162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677567 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711442 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677567 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711442 - -
          mouse-6 v1.1 (Illumina) scl0002692.1_59-S - -
          mouse-6 v1.1 (Illumina) scl25528.7.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550075 - -
          Illumina Sentrix mouse-6.v1 bead array 3710162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002692.1_59-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25528.7.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550075 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677567 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711442 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711442 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677567 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002692.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25528.7.1_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline