| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500005787.mm.1 |
TC0500000652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005787.mm.1 |
NR_015582.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005787.mm.1 |
NR_110363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005789.mm.1 |
TC0500000652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005789.mm.1 |
NR_110363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005790.mm.1 |
TC0500000652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005790.mm.1 |
NR_110363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500005792.mm.1 |
TC0500000652.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500005792.mm.1 |
NR_110363.1 |
Refseq |