Search
Search

Wdr95| Gene Symbol: Wdr95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029440.3 | NP_083716.2 | 9 - 10 | 853 | 70 | |
| GenBank mRNA | AK015309.1 | - | 7 - 8 | 852 | 70 | |
| AK087389.1 | - | 9 - 10 | 853 | 70 | ||
Set Membership: |
|