Search
Search
| Gene Symbol: Ubl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011908.2 | NP_036038.1 | 2 - 3 | 942 | 58 | |
| GenBank mRNA | AF044222.1 | - | 2 - 3 | 884 | 58 | |
| AF044223.1 | - | 2 - 3 | 203 | 58 | ||
| AK134598.1 | - | 2 - 3 | 310 | 58 | ||
| AK145952.1 | - | 2 - 3 | 621 | 58 | ||
| BC024507.1 | - | 4 - 5 | 792 | 4983604 | 58 | |
| BC025595.1 | - | 2 - 3 | 597 | 5342735 | 58 | |
| BC043729.1 | - | 4 - 5 | 792 | 4983604 | 58 | |
Set Membership: |
|