Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219152_m1
          See other 1700094D03RIK GE Assays ›
          Gene Symbol:
          1700094D03Rik
          Gene Name:
          RIKEN cDNA 1700094D03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 90062796 - 90068347 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219152_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700094D03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028567.1 NP_082843.1 3 - 4 255 105
          GenBank mRNA AK007060.1 - 3 - 4 256 105
          BC048560.1 - 3 - 4 251 6703545 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000775.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 NM_028567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 AK007060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 BC048560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 NM_028567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 AK007060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308446 BC048560.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156282 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146937 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 NM_028567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 AK007060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 BC048560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 NM_028567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 AK007060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308446 BC048560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 NM_028567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 AK007060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 BC048560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 NM_028567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 AK007060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308446 BC048560.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493625 - -
          Affymetrix HT MG-430 PM Array Plate 1447556_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1429907_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493626 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493612 - -
          Mouse Genome 430 2.0 Array 1429907_at - -
          Mouse Genome 430 2.0 Array 1443955_at - -
          Mouse Genome 430 2.0 Array 1447556_x_at - -
          Mouse Genome 430 2.0 Array 1429907_PM_at - -
          Mouse Genome 430 2.0 Array 1443955_PM_at - -
          Mouse Genome 430 2.0 Array 1447556_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73545_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73545_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300005962.mm.1 NM_028567.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005962.mm.1 NM_028567.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005962.mm.1 TC0300000775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005961.mm.1 NM_028567.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005961.mm.1 NM_028567.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005961.mm.1 TC0300000775.mm.1 MTA TC
          Mouse Expression Set 430 1429907_PM_at - -
          Mouse Expression Set 430 1429907_at - -
          Mouse Expression Set 430 1443955_at - -
          Mouse Expression Set 430 1447556_x_at - -
          Mouse Expression Set 430 1443955_PM_at - -
          Mouse Expression Set 430 1447556_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1429907_at - -
          Mouse Genome 430A 2.0 Array 1429907_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429907_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429907_at - -
          Affymetrix HT Mouse Genome 430B Array 1447556_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443955_at - -
          Affymetrix HT Mouse Genome 430B Array 1443955_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447556_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359219 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259302 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259302 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0