| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22329268 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2163238 |
NM_207247.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2163238 |
AY518698.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2163238 |
BC128291.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529358 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269319 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
113829 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667157 |
AK080547.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826438 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
950506 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449409 |
AK029811.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449409 |
BC030867.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449409 |
BC085317.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449409 |
BC108338.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449409 |
NM_153544.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113290 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81344 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135030 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2163238 |
NM_207247.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2163238 |
AY518698.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2163238 |
BC128291.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329229 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329223 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329225 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329216 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329242 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329232 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329227 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329208 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329248 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329221 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329220 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329243 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329245 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329213 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329219 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329215 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329231 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329238 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329218 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329240 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329233 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329236 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22329217 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135030 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04988304 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04988289 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P427607 |
NM_207247.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122215 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135030 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02328254 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03163095 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03163093 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03163091 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02328270 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22329268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329267 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329314 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329321 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329319 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329344 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329262 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329290 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329383 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329332 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329295 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329323 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329258 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329233 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329308 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329304 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329275 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329342 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329293 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329287 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329376 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329251 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329298 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329307 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329336 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329238 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329255 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329371 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329281 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329365 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329236 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329309 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329325 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329350 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329277 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329368 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329329 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329340 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329352 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329363 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329257 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329300 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22329348 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329314 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329281 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329311 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329216 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329293 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329306 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329238 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329254 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329251 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329321 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329225 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329328 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329267 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329298 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329242 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329245 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329292 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329277 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02304093 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329240 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329233 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329208 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329266 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329283 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329246 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02304095 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329304 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03102796 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329319 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329264 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329274 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329303 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329261 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329255 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329285 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02906587 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329295 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329217 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329215 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329258 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329268 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329256 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329220 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329279 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329317 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02297872 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329227 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329219 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329300 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329330 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329236 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329307 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329243 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329247 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329275 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329231 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329290 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02328258 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329309 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329262 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02545251 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329310 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329323 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22329223 |
- |
- |