Search
Search

Crtc2| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028881.2 | NP_083157.1 | 1 - 2 | 281 | 71 | |
| GenBank mRNA | AK079382.1 | - | 1 - 2 | 239 | 71 | |
| AK156192.1 | - | 1 - 2 | 260 | 71 | ||
| AK172084.1 | - | 1 - 2 | 239 | 71 | ||
| BC023831.1 | - | 1 - 2 | 276 | 5345301 | 71 | |
| BC032183.1 | - | 1 - 2 | 286 | 5310152 | 71 | |
| BC033300.1 | - | 1 - 2 | 276 | 4166411 | 71 | |
| BU612513.1 | - | 1 - 2 | 222 | 71 | ||
| BY085181.1 | - | 1 - 2 | 258 | 71 | ||
Set Membership: |
|