Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219987_m1
          See other NUP210L GE Assays ›
          Gene Symbol:
          Nup210l
          Gene Name:
          nucleoporin 210-like
          Gene Aliases:
          4930548O11Rik, R26-EGFP, Tg(Gt(ROSA)26Sor-EGFP)130910Eps
          Chromosome Location:
          Chr.3: 90101569 - 90212151 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219987_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup210l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029937.1 NP_084213.1 24 - 25 3327 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000780.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          Sertoli cell development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P192059 NM_029937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P192059 NM_029937.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK032447.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK044754.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK046401.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK051096.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK122299.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK133157.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 AK172108.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 BC027126.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597728 NM_172286.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808673 AK054208.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300596 AK033559.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300596 AK076991.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300596 BC060213.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300596 BC108346.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300596 NM_178896.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51060 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68162 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150604 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143226 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P192059 NM_029937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P192059 NM_029937.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04985585 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143226 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00697596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04985700 - -
          Whole Mouse Genome G4122A A_52_P884807 NM_029937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P884807 AK019752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P192059 NM_029937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P192059 NM_029937.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04985700 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143226 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03171751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02291237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02851053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02581041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03060859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02403933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03060860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02738726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03030446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03111622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02339748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323794 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430909_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493670 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493688 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493660 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493664 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493662 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493640 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493666 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493690 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493668 - -
          Mouse Genome 430 2.0 Array 1430909_at - -
          Mouse Genome 430 2.0 Array 1430909_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77595_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77595_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006043.mm.1 NM_029937.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006043.mm.1 NM_029937.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006043.mm.1 TC0300000780.mm.1 MTA TC
          Mouse Expression Set 430 1430909_PM_at - -
          Mouse Expression Set 430 1430909_at - -
          Affymetrix HT Mouse Genome 430B Array 1430909_at - -
          Affymetrix HT Mouse Genome 430B Array 1430909_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 768690 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231456 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0