Search
Search

Samd10| Gene Symbol: Samd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172676.2 | NP_766264.2 | 1 - 2 | 211 | 113 | |
| GenBank mRNA | AK171151.1 | - | 1 - 2 | 217 | 113 | |
| BC052921.1 | - | 1 - 2 | 211 | 6311715 | 113 | |
Set Membership: |
|