Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220420_m1
          See other RTEL1 GE Assays ›
          Gene Symbol:
          Rtel1
          Gene Name:
          regulator of telomere elongation helicase 1
          Gene Aliases:
          AI451565, AW540478, Rtel
          Chromosome Location:
          Chr.2: 181288028 - 181356616 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220420_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtel1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001882.3 NP_001001882.3 16 - 17 1630 63
          NM_001166665.1 NP_001160137.1 16 - 17 1630 63
          NM_001166666.1 NP_001160138.1 16 - 17 1630 63
          NM_001166667.1 NP_001160139.1 16 - 17 1630 63
          NM_001166668.1 NP_001160140.1 16 - 17 1630 63
          NR_030710.1 15 - 16 1497 63
          GenBank mRNA AK142225.1 - 16 - 17 1617 63
          AK173097.1 - 16 - 17 1592 63
          AK184857.1 - 2 - 3 135 63
          AY481619.1 - 16 - 17 1569 63
          AY481620.1 - 16 - 17 1569 63
          AY481621.1 - 16 - 17 1569 63
          AY481622.1 - 16 - 17 1569 63
          AY481623.1 - 16 - 17 1569 63
          BC031201.1 - 10 - 11 795 63
          BC057352.1 - 16 - 17 1602 63
          BC105578.1 - 16 - 17 1542 30478629 63
          BC144977.1 - 15 - 16 1382 9053525 63
          BC144978.1 - 15 - 16 1382 9053526 63
          BC145658.1 - 15 - 16 1382 40130961 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002821.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtel1

          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          nucleobase-containing compound metabolic process
          DNA repair
          cellular response to DNA damage stimulus
          regulation of double-strand break repair via homologous recombination
          DNA duplex unwinding
          nucleotide binding
          nucleic acid binding
          DNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          ATP binding
          ATP-dependent helicase activity
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001001882.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001001882.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166665.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166665.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166666.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166666.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166667.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166667.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166668.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NM_001166668.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NR_030710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 NR_030710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023176.mm.1 TC0200002821.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline