Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220470_m1
          See other SUPT7L GE Assays ›
          Gene Symbol:
          Supt7l
          Gene Name:
          suppressor of Ty 7-like
          Gene Aliases:
          2610524B01Rik, 6030455L14Rik
          Chromosome Location:
          Chr.5: 31514569 - 31526762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220470_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Supt7l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028150.2 NP_082426.1 4 - 5 1314 79
          GenBank mRNA AK012134.1 - 4 - 5 1260 79
          AK144989.1 - 4 - 5 1186 79
          BC031447.1 - 4 - 5 1237 4502314 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002186.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Supt7l

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3 acetylation
          maintenance of protein location in nucleus
          regulation of nucleic acid-templated transcription
          transcription coactivator activity
          histone acetyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23578562 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 AK144989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 BC031447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 AK012134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 AK144989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 BC031447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 AK012134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13226 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178157 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 AK144989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 BC031447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 AK012134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 NM_028150.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 AK144989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 BC031447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 AK012134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 AK144989.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 BC031447.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 AK012134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 AK144989.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 BC031447.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 AK012134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23578540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178157 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180301 - -
          Whole Mouse Genome G4122A A_52_P241544 AK144989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 BC031447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 AK012134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 NM_028150.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 AK144989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 BC031447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 AK012134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P241544 NM_028150.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178157 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04699425 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04699431 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04699415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04699420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04699423 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23578562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23578502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04455626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04455625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04452100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578525 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529223 - -
          Affymetrix HT MG-430 PM Array Plate 1429876_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433373_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433374_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529218 - -
          Mouse Genome 430 2.0 Array 1433374_at - -
          Mouse Genome 430 2.0 Array 1433373_PM_at - -
          Mouse Genome 430 2.0 Array 1433373_at - -
          Mouse Genome 430 2.0 Array 1429876_at - -
          Mouse Genome 430 2.0 Array 1429876_PM_at - -
          Mouse Genome 430 2.0 Array 1433374_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72195_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72195_at - -
          Affymetrix Mouse Clariom S Array PSR0500020020.mm.1 TC0500002186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020020.mm.1 NM_028150.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020020.mm.1 NM_028150.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020021.mm.1 TC0500002186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020021.mm.1 NM_028150.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020021.mm.1 NM_028150.2 Refseq
          Mouse Expression Set 430 1429876_PM_at - -
          Mouse Expression Set 430 1429876_at - -
          Affymetrix HT Mouse Genome 430B Array 1433373_at - -
          Affymetrix HT Mouse Genome 430B Array 1433374_at - -
          Affymetrix HT Mouse Genome 430B Array 1433374_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429876_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433373_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429876_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 718621 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747698 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0