Search
Search

Ift172| Gene Symbol: Ift172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026298.5 | NP_080574.5 | 18 - 19 | 2037 | 58 | |
| GenBank mRNA | AK043642.1 | - | 18 - 19 | 1985 | 58 | |
| AK122461.1 | - | 3 - 4 | 331 | 58 | ||
| AY339616.1 | - | 18 - 19 | 1973 | 58 | ||
| BC042583.1 | - | 18 - 19 | 1976 | 58 | ||
| BC066096.1 | - | 18 - 19 | 2030 | 6856438 | 58 | |
Set Membership: |
|