Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220560_g1
          See other SLC5A6 GE Assays ›
          Gene Symbol:
          Slc5a6
          Gene Name:
          solute carrier family 5 (sodium-dependent vitamin transporter), member 6
          Gene Aliases:
          E430023I20
          Chromosome Location:
          Chr.5: 31036036 - 31048924 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220560_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc5a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177621.1 NP_001171092.1 17 - 18 2517 70
          NM_001177622.1 NP_001171093.1 17 - 18 2299 70
          NM_177870.5 NP_808538.1 16 - 17 2406 70
          GenBank mRNA AK088682.1 - 17 - 18 2205 70
          AK158896.1 - 17 - 18 2518 70
          AK161633.1 - 16 - 17 2330 70
          AY572835.1 - 14 - 15 1757 70
          BC065061.1 - 17 - 18 2298 70
          BC085132.1 - 16 - 17 2102 6416422 70
          BC117852.1 - 16 - 17 2059 40094012 70
          BC117853.1 - 16 - 17 2059 40094013 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc5a6

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sodium ion transport
          biotin transport
          pantothenate transmembrane transport
          sodium ion transmembrane transport
          transmembrane transport
          transporter activity
          sodium-dependent multivitamin transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019736.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019723.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019727.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019729.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019737.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019748.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019748.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019748.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019731.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019739.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019742.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019743.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019751.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019751.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019751.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019724.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019730.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019732.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019734.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019735.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019745.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019746.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019740.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019749.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019749.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019749.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019753.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019753.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019753.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019722.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019725.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019726.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019733.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019741.mm.1 NM_177870.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019752.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019752.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019752.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 TC0500002170.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_001177621.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_001177621.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_001177622.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_001177622.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_177870.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019738.mm.1 NM_177870.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 711199 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline