Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01221140_m1
          See other SRSF12 GE Assays ›
          Gene Symbol:
          Srsf12
          Gene Name:
          serine/arginine-rich splicing factor 12
          Gene Aliases:
          B830039L16, Sfrs13b, Srrp, Srsf13b
          Chromosome Location:
          Chr.4: 33208987 - 33233344 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01221140_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srsf12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177774.4 NP_808442.2 2 - 3 525 112
          GenBank mRNA AK046869.1 - 2 - 3 526 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000282.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srsf12

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825735 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825737 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825736 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230347 NM_177774.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230347 AK046869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230347 NM_177774.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230347 AK046869.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753332 AK083224.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753332 AK147451.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753332 BC060736.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753332 NM_001081377.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11237 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116108 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141938 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230347 NM_177774.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230347 AK046869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230347 NM_177774.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230347 AK046869.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825751 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116108 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05196705 - -
          Whole Mouse Genome G4122A A_51_P230347 NM_177774.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230347 AK046869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230347 NM_177774.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230347 AK046869.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119070 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116108 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03857649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03882315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03882310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03882311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03882313 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825735 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825737 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22825736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03527061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03890488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03857648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04028355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03882311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04070862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03657113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03860449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03640718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03640722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04025241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03620175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03687230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04066490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22825751 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10503881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503880 - -
          Affymetrix HT MG-430 PM Array Plate 1437168_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10503876 - -
          Mouse Genome 430 2.0 Array 1437168_PM_at - -
          Mouse Genome 430 2.0 Array 1437168_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 272009_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 272009_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400001845.mm.1 NM_177774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001845.mm.1 NM_177774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001845.mm.1 TC0400000282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400001846.mm.1 NM_177774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001846.mm.1 NM_177774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001846.mm.1 TC0400000282.mm.1 MTA TC
          Mouse Expression Set 430 1437168_PM_at - -
          Mouse Expression Set 430 1437168_at - -
          Affymetrix HT Mouse Genome 430B Array 1437168_at - -
          Affymetrix HT Mouse Genome 430B Array 1437168_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339692 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060332 - -
          Illumina mouse-6 v1.0 expression beadchip 3060332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2633563 - -
          mouse-6 v1.1 (Illumina) scl25570.5.1_79-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25570.5.1_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2953888 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2953888 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25570.5.1_79-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline