Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01221891_m1
          See other 1810011H11RIK GE Assays ›
          Gene Symbol:
          1810011H11Rik
          Gene Name:
          RIKEN cDNA 1810011H11 gene
          Gene Aliases:
          4733401I05Rik
          Chromosome Location:
          Chr.14: 32785885 - 32817969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01221891_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810011H11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163616.1 NP_001157088.1 3 - 4 321 61
          GenBank mRNA AK007434.1 - 3 - 4 322 61
          AK014636.1 - 2 - 3 262 61

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK014636.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 AK007434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 AK014636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 NM_001163616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 AK007434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 AK014636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P308413 NM_001163616.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178480 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 AK007434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 AK014636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 NM_001163616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 AK007434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 AK014636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P308413 NM_001163616.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178480 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08076369 - -
          Whole Mouse Genome G4122A A_52_P308413 AK007434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308413 AK014636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308413 NM_001163616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308413 AK007434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308413 AK014636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308413 NM_001163616.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11130995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11680275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11675069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11414090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11202941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11130982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11694349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11439102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11229064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11306459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11130989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11134592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11421855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11202931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11680277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11306460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11134594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11130984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11130987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11781105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11667645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11234434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11675068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29520009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11234431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29519955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10413929 - -
          Affymetrix HT MG-430 PM Array Plate 1454122_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429604_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10413928 - -
          Mouse Genome 430 2.0 Array 1454122_PM_at - -
          Mouse Genome 430 2.0 Array 1429604_PM_at - -
          Mouse Genome 430 2.0 Array 1454122_at - -
          Mouse Genome 430 2.0 Array 1429604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69069_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69069_at - -
          Affymetrix Mouse Clariom S Array PSR1400003113.mm.1 NM_001163616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003113.mm.1 NM_001163616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003113.mm.1 TC1400000350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400003114.mm.1 NM_001163616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003114.mm.1 NM_001163616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003114.mm.1 TC1400000350.mm.1 MTA TC
          Mouse Expression Set 430 1429604_at - -
          Mouse Expression Set 430 1429604_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429604_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429604_at - -
          Affymetrix HT Mouse Genome 430B Array 1454122_at - -
          Affymetrix HT Mouse Genome 430B Array 1454122_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 489931 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103450050 - -
          MouseWG-6 v2 Expression BeadChip 105360066 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2500158 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259018 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222269 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0