Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222827_m1
          See other S100A1 GE Assays ›
          Gene Symbol:
          S100a1
          Gene Name:
          S100 calcium binding protein A1
          Gene Aliases:
          AI266795, S100, S100a
          Chromosome Location:
          Chr.3: 90511034 - 90514330 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222827_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011309.3 NP_035439.1 2 - 3 278 68
          GenBank mRNA AF087687.1 - 2 - 3 245 68
          AK002721.1 - 2 - 3 219 68
          AK012578.1 - 2 - 3 278 68
          AK017279.1 - 2 - 3 214 68
          AK168822.1 - 2 - 3 214 68
          BC005590.1 - 2 - 3 350 3964547 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002374.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100a1

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of heart contraction
          positive regulation of voltage-gated calcium channel activity
          calcium ion binding
          protein binding
          identical protein binding
          protein homodimerization activity
          S100 protein binding
          metal ion binding
          calcium-dependent protein binding
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979650 NM_011309.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979650 AK012578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979650 AK168822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979650 BC005590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979645 NM_011309.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979645 AF087687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979645 AK002721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979645 AK012578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979645 AK017279.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765622 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148979 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979645 NM_011309.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979645 AF087687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979645 AK002721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979645 AK012578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979645 AK017279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979650 NM_011309.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979650 AK012578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979650 AK168822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979650 BC005590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172475 NM_011309.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172475 AF087687.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172475 AK002721.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172475 AK012578.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172475 AK017279.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22325669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148979 - -
          Whole Mouse Genome G4122A A_51_P172475 NM_011309.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172475 AF087687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172475 AK002721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172475 AK012578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172475 AK017279.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134524 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02532070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22325676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22325656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02532076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499857 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499856 - -
          Affymetrix HT MG-430 PM Array Plate 1417421_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419814_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499854 - -
          Mouse Genome 430 2.0 Array 1419814_s_at - -
          Mouse Genome 430 2.0 Array 1417421_at - -
          Mouse Genome 430 2.0 Array 1419814_PM_s_at - -
          Mouse Genome 430 2.0 Array 1417421_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20193_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20193_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300018508.mm.1 NM_011309.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018508.mm.1 NM_011309.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018508.mm.1 TC0300002374.mm.1 MTA TC
          Mouse Expression Set 430 1417421_PM_at - -
          Mouse Expression Set 430 1417421_at - -
          Murine Genome U74v2 Set 95453_PM_f_at - -
          Murine Genome U74v2 Set 95452_i_at - -
          Murine Genome U74v2 Set 95452_PM_i_at - -
          Murine Genome U74v2 Set 95453_f_at - -
          Mouse Genome 430A 2.0 Array 1417421_PM_at - -
          Mouse Genome 430A 2.0 Array 1417421_at - -
          Affymetrix HT Mouse Genome 430A Array 1417421_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419814_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417421_at - -
          Affymetrix HT Mouse Genome 430A Array 1419814_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919747 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770707 - -
          Illumina mouse-6 v1.0 expression beadchip 6770707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225602 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225602 - -
          mouse-6 v1.1 (Illumina) scl21916.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21916.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225602 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225602 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21916.1_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0