Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01223417_m1
          See other PAQR6 GE Assays ›
          Gene Symbol:
          Paqr6
          Gene Name:
          progestin and adipoQ receptor family member VI
          Gene Aliases:
          1500001B10Rik
          Chromosome Location:
          Chr.3: 88364072 - 88368541 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01223417_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Paqr6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198410.3 NP_940802.1 5 - 6 568 90
          GenBank mRNA AK139144.1 - 5 - 6 569 90
          AY424295.1 - 5 - 6 569 90
          BC120512.1 - 4 - 5 531 40129435 90
          BC125428.1 - 4 - 5 531 40129943 90
          BC144954.1 - 3 - 4 606 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000725.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Paqr6

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313386 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313385 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC120512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC125428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC144954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 AK139144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC030379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC120512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC125428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC144954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 AK139144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475628 BC030379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148655 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971025 AK139144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971025 BC030379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971025 AK005086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971025 NM_198410.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971025 AI607833.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75788 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173075 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971025 AK139144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971025 BC030379.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971025 AK005086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971025 NM_198410.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971025 AI607833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148655 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC120512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC125428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC144954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 AK139144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC030379.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC120512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC125428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC144954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 AK139144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475628 BC030379.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22313383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04980734 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173075 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04980759 - -
          Whole Mouse Genome G4122A A_51_P475628 BC120512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC125428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC144954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 AK139144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC030379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC120512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC125428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC144954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 AK139144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475628 BC030379.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127581 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106574 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04980734 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296742 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296732 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296726 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02296739 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22313386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02732546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03098688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03098686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493241 - -
          Affymetrix HT MG-430 PM Array Plate 1455216_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493235 - -
          Mouse Genome 430 2.0 Array 1455216_at - -
          Mouse Genome 430 2.0 Array 1455216_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68957_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68957_at - -
          Affymetrix Mouse Clariom S Array PSR0300005141.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005141.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005141.mm.1 TC0300000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005137.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005137.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005137.mm.1 TC0300000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005138.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005138.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005138.mm.1 TC0300000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005139.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005139.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005139.mm.1 TC0300000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005136.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005136.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005136.mm.1 TC0300000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005140.mm.1 NM_198410.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005140.mm.1 NM_198410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005140.mm.1 TC0300000725.mm.1 MTA TC
          Mouse Expression Set 430 1455216_PM_at - -
          Mouse Expression Set 430 1455216_at - -
          Murine Genome U74v2 Set 170661_PM_i_at - -
          Murine Genome U74v2 Set 170661_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1455216_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455216_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 740078 - -
          ABI Mouse Genome Survey Microarray 818656 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550341 - -
          MouseWG-6 v2 Expression BeadChip 2630093 - -
          MouseWG-6 v2 Expression BeadChip 580450 - -
          Illumina mouse-6 v1.0 expression beadchip 2630093 - -
          Illumina mouse-6 v1.0 expression beadchip 5550341 - -
          Illumina mouse-6 v1.0 expression beadchip 580450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221469 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241105 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241105 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221469 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727184 - -
          mouse-6 v1.1 (Illumina) scl23001.2_10-S - -
          mouse-6 v1.1 (Illumina) scl0068957.1_272-S - -
          mouse-6 v1.1 (Illumina) scl068957.3_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550341 - -
          Illumina Sentrix mouse-6.v1 bead array 580450 - -
          Illumina Sentrix mouse-6.v1 bead array 2630093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068957.3_51-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23001.2_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0068957.1_272-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221469 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2727184 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2727184 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221469 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068957.3_51-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0