Search
Search

1700011E24Rik| Gene Symbol: 1700011E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029298.1 | NP_083574.1 | 3 - 4 | 479 | 59 | |
| GenBank mRNA | AK005861.1 | - | 3 - 4 | 424 | 59 | |
| BE456690.1 | - | 4 - 5 | 59 | |||
Set Membership: |
|