Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01223848_m1
          See other FBXL17 GE Assays ›
          Gene Symbol:
          Fbxl17
          Gene Name:
          F-box and leucine-rich repeat protein 17
          Gene Aliases:
          6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13
          Chromosome Location:
          Chr.17: 63045949 - 63504042 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01223848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxl17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015794.1 NP_056609.1 8 - 9 2526 63
          GenBank mRNA AF176529.1 - 8 - 9 1246 63
          AK004419.1 - 2 - 3 208 63
          AK154497.1 - 8 - 9 1035 63
          BC058120.1 - 5 - 6 584 6810624 63
          BC138330.1 - 5 - 6 562 8861350 63
          BC138331.1 - 5 - 6 562 8861351 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002384.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700022245.mm.1 NM_015794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022245.mm.1 NM_015794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022245.mm.1 TC1700002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700022246.mm.1 NM_015794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022246.mm.1 NM_015794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022246.mm.1 TC1700002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700022249.mm.1 NM_015794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022249.mm.1 NM_015794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022249.mm.1 TC1700002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700022247.mm.1 NM_015794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022247.mm.1 NM_015794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022247.mm.1 TC1700002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700022248.mm.1 NM_015794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022248.mm.1 NM_015794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700022248.mm.1 TC1700002384.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0