Search
Search
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031848.1 | - | 1 - 2 | 134 | 68 | |
| AK036017.1 | - | 1 - 2 | 82 | 68 | ||
| AK041354.1 | - | 1 - 2 | 111 | 68 | ||
| AK042540.1 | - | 1 - 2 | 76 | 68 | ||
| AK163870.1 | - | 1 - 2 | 83 | 68 | ||
| AY753561.1 | - | 1 - 2 | 38 | 68 | ||
| AY753562.1 | - | 1 - 2 | 38 | 68 | ||
| BC056428.1 | - | 1 - 2 | 69 | 5716245 | 68 | |
| DQ025533.1 | - | 1 - 2 | 38 | 68 | ||
Set Membership: |
|