Search
Search
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201529.2 | NP_963287.2 | 2 - 3 | 369 | 64 | |
| GenBank mRNA | BB614044.1 | - | 2 - 3 | 385 | 64 | |
| EF071857.1 | - | 2 - 3 | 340 | 64 | ||
Set Membership: |
|