Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01224217_m1
          See other COMMD6 GE Assays ›
          Gene Symbol:
          Commd6
          Gene Name:
          COMM domain containing 6
          Gene Aliases:
          1110059J08Rik, 1700063H17Rik
          Chromosome Location:
          Chr.14: 101633766 - 101640471 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01224217_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Commd6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033132.3 NP_001028304.1 3 - 4 242 68
          NM_001168592.1 NP_001162064.1 2 - 3 230 68
          GenBank mRNA AK004316.1 - 3 - 4 216 68
          AK008220.1 - 3 - 4 224 68
          AK009796.1 - 3 - 4 237 68
          AK011226.1 - 3 - 4 227 68
          AK018711.1 - 3 - 4 216 68
          AK019351.1 - 3 - 4 218 68
          AK019432.1 - 3 - 4 219 68
          AK020234.1 - 3 - 4 215 68
          AK034223.1 - 1 - 2 3490 68
          AK045407.1 - 2716 68
          BC132261.1 - 3 - 4 229 40130538 68
          BC132265.1 - 3 - 4 229 40130542 68
          BC145360.1 - 3 - 4 229 9053918 68
          BC145361.1 - 2 - 3 217 9053919 68
          CK621880.1 - 2 - 3 193 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002625.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of NF-kappaB transcription factor activity
          NF-kappaB binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856056 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856055 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 CK128085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 CK621880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 AK034223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 AK045407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 BC132261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 CK128085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 CK621880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 AK034223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 AK045407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P446120 BC132261.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731089 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152775 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 CK128085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 CK621880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 AK034223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 AK045407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 BC132261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 CK128085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 CK621880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 AK034223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 AK045407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P446120 BC132261.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29856092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29856091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29856094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29856089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08218323 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152775 - -
          Whole Mouse Genome G4122A A_52_P446120 CK128085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 CK621880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 AK034223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 AK045407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 BC132261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 CK128085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 CK621880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 AK034223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 AK045407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446120 BC132261.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119360 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152775 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856056 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29856058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29855991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29856036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11202360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11696096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11814936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11824531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11673719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11802438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29856087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10422054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10422053 - -
          Affymetrix HT MG-430 PM Array Plate 1435863_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10422052 - -
          Mouse Genome 430 2.0 Array 1435863_PM_at - -
          Mouse Genome 430 2.0 Array 1435863_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66200_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66200_at - -
          Affymetrix Mouse Clariom S Array PSR1400019595.mm.1 NM_001033132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019595.mm.1 NM_001033132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019595.mm.1 NM_001168592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019595.mm.1 NM_001168592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019595.mm.1 TC1400002625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019600.mm.1 NM_001033132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019600.mm.1 NM_001033132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019600.mm.1 NM_001168592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019600.mm.1 NM_001168592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019600.mm.1 TC1400002625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019594.mm.1 NM_001033132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019594.mm.1 NM_001033132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019594.mm.1 NM_001168592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019594.mm.1 NM_001168592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019594.mm.1 TC1400002625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400019596.mm.1 NM_001033132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019596.mm.1 NM_001033132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019596.mm.1 NM_001168592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019596.mm.1 NM_001168592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400019596.mm.1 TC1400002625.mm.1 MTA TC
          Mouse Expression Set 430 1435863_at - -
          Mouse Expression Set 430 1435863_PM_at - -
          Murine Genome U74v2 Set 96881_at - -
          Murine Genome U74v2 Set 96881_PM_at - -
          Murine Genome U74v2 Set 166932_at - -
          Murine Genome U74v2 Set 166932_PM_at - -
          Mouse Genome 430A 2.0 Array 1435863_at - -
          Mouse Genome 430A 2.0 Array 1435863_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435863_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435863_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 915242 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160703 - -
          mouse-6 v1.1 (Illumina) GI_84370277-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2993720 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993720 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_84370277-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline