Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01225747_m1
          See other RIMS1 GE Assays ›
          Gene Symbol:
          Rims1
          Gene Name:
          regulating synaptic membrane exocytosis 1
          Gene Aliases:
          C030033M19Rik, RIM1, RIM1a, RIM1alpha, Rab3ip1, Rim, Serg1, mKIAA0340
          Chromosome Location:
          Chr.1: 22288422 - 22432843 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01225747_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rims1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012623.1 NP_001012641.1 1247 63
          NM_001012624.1 NP_001012642.1 1247 63
          NM_001012625.1 NP_001012643.1 1247 63
          NM_053270.1 NP_444500.1 1247 63
          NM_183018.2 NP_898839.2 1247 63
          GenBank mRNA AK134597.1 - 158 63
          AK162935.1 - 2 - 3 143 63
          AK220335.1 - 1180 63
          AY356534.1 - 1247 63
          AY356535.1 - 1247 63
          AY356536.1 - 1247 63
          AY356537.1 - 1247 63
          AY356538.1 - 1247 63

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK134597.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rims1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          transport
          neurotransmitter transport
          intracellular protein transport
          exocytosis
          positive regulation of gene expression
          positive regulation of synaptic vesicle priming
          regulation of membrane potential
          regulation of long-term neuronal synaptic plasticity
          calcium ion-regulated exocytosis of neurotransmitter
          long-term synaptic potentiation
          spontaneous neurotransmitter secretion
          positive regulation of inhibitory postsynaptic potential
          positive regulation of dendrite extension
          regulation of synaptic vesicle exocytosis
          positive regulation of excitatory postsynaptic potential
          protein binding
          SH3 domain binding
          Rab GTPase binding
          protein kinase binding
          ion channel binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100016749.mm.1 NM_001012623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016749.mm.1 NM_001012624.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016749.mm.1 NM_001012625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016749.mm.1 TC0100002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016747.mm.1 NM_001012623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016747.mm.1 NM_001012624.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016747.mm.1 NM_001012625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016747.mm.1 TC0100002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016725.mm.1 NM_053270.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016725.mm.1 NM_183018.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016725.mm.1 TC0100002054.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016746.mm.1 NM_001012623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016746.mm.1 NM_001012624.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016746.mm.1 NM_001012625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016746.mm.1 TC0100002054.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 301377 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0