Search
Search

Vps54| Gene Symbol: Vps54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290628.1 | NP_001277557.1 | 6 - 7 | 952 | 76 | |
| NM_139061.5 | NP_620692.3 | 6 - 7 | 988 | 76 | ||
| GenBank mRNA | AF424699.2 | - | 6 - 7 | 739 | 76 | |
| AK077306.1 | - | 4 - 5 | 370 | 76 | ||
| BC025012.2 | - | 6 - 7 | 663 | 5042879 | 76 | |
Set Membership: |
|