Search
Search

Rbm26| Gene Symbol: Rbm26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134077.4 | NP_598838.3 | 6 - 7 | 1365 | 68 | |
| GenBank mRNA | AK049083.1 | - | 5 - 6 | 821 | 68 | |
| AK051686.1 | - | 6 - 7 | 1341 | 68 | ||
| AK139077.1 | - | 6 - 7 | 1366 | 68 | ||
| AK172039.1 | - | 6 - 7 | 1344 | 68 | ||
| BC049360.1 | - | 6 - 7 | 1341 | 68 | ||
| BC066051.1 | - | 6 - 7 | 1302 | 68 | ||
Set Membership: |
|