| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000022025.mm.1 |
NM_001033333.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022025.mm.1 |
NM_001033333.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022025.mm.1 |
TC1000002915.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000022026.mm.1 |
NM_001033333.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022026.mm.1 |
NM_001033333.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022026.mm.1 |
TC1000002915.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000022027.mm.1 |
NM_001033333.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022027.mm.1 |
NM_001033333.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022027.mm.1 |
TC1000002915.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000022028.mm.1 |
NM_001033333.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022028.mm.1 |
NM_001033333.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022028.mm.1 |
TC1000002915.mm.1 |
MTA TC |