Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227700_m1
          See other DLGAP1 GE Assays ›
          Gene Symbol:
          Dlgap1
          Gene Name:
          discs, large (Drosophila) homolog-associated protein 1
          Gene Aliases:
          4933422O14Rik, 9630002F18, AI845682, AI848168, BB075781, D17Bwg0511e, GKAP/SAPAP, GKPA/SAPAP, Gkap, Sapap1, mKIAA4162
          Chromosome Location:
          Chr.17: 69969078 - 70821413 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227700_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlgap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128180.1 NP_001121652.1 2 - 3 369 82
          NM_001128181.1 NP_001121653.1 2 - 3 341 82
          NM_027712.3 NP_081988.3 3 - 4 1356 82
          NM_177639.6 NP_808307.2 3 - 4 1356 82
          GenBank mRNA AK031410.1 - 7 - 8 2046 82
          AK035763.1 - 6 - 7 1907 82
          AK136926.1 - 2 - 3 302 82
          AK147448.1 - 7 - 8 2173 82
          AK163430.1 - 2 - 3 538 82
          AK163689.1 - 2 - 3 350 82
          AK220515.1 - 6 - 7 2236 82
          BC062120.1 - 3 - 4 1357 6822047 82
          BC094369.1 - 2 - 3 565 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001113.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dlgap1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          signaling
          protein localization to synapse
          regulation of proteasomal protein catabolic process
          aggresome assembly
          protein domain specific binding
          protein complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700010495.mm.1 NM_001128181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010495.mm.1 NM_001128181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010495.mm.1 TC1700001113.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010488.mm.1 NM_001128180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010488.mm.1 NM_001128180.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010488.mm.1 TC1700001113.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010494.mm.1 NM_001128181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010494.mm.1 NM_001128181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010494.mm.1 TC1700001113.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010491.mm.1 NM_001128180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010491.mm.1 NM_001128180.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010491.mm.1 TC1700001113.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_001128180.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_001128180.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_001128181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_001128181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_027712.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_027712.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_177639.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 NM_177639.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010499.mm.1 TC1700001113.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0