Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227863_m1
          See other 2010003K11RIK GE Assays ›
          Gene Symbol:
          2010003K11Rik
          Gene Name:
          RIKEN cDNA 2010003K11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 4496788 - 4498583 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227863_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010003K11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027237.1 NP_081513.1 1 - 2 343 57
          GenBank mRNA AK008072.1 - 1 - 2 321 57
          BC021951.1 - 1 - 2 343 5102159 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000924.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2010003K11Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 NM_027237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 AK008072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 BC021951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 NM_027237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 AK008072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450278 BC021951.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142280 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 NM_027237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 AK008072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 BC021951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 NM_027237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 AK008072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450278 BC021951.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31923454 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142280 - -
          Whole Mouse Genome G4122A A_51_P450278 NM_027237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450278 AK008072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450278 BC021951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450278 NM_027237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450278 AK008072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450278 BC021951.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14236652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14236655 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14236653 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14236649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31923413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14300978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14333377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14262504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14423823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14307141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14391410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14179085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14175135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14151299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14434933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14154864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14154865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14307144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14196836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31923434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10464773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10464774 - -
          Affymetrix HT MG-430 PM Array Plate 1424626_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10464772 - -
          Mouse Genome 430 2.0 Array 1424626_at - -
          Mouse Genome 430 2.0 Array 1424626_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69861_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69861_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900008132.mm.1 NM_027237.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008132.mm.1 NM_027237.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008132.mm.1 TC1900000924.mm.1 MTA TC
          Mouse Expression Set 430 1424626_PM_at - -
          Mouse Expression Set 430 1424626_at - -
          Mouse Genome 430A 2.0 Array 1424626_at - -
          Mouse Genome 430A 2.0 Array 1424626_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424626_at - -
          Affymetrix HT Mouse Genome 430A Array 1424626_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617328 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0