Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01227912_s1
          See other MALAT1 GE Assays ›
          Gene Symbol:
          Malat1
          Gene Name:
          metastasis associated lung adenocarcinoma transcript 1 (non-coding RNA)
          Gene Aliases:
          2210401K01Rik, 9430072K23Rik, AI647968, Neat2
          Chromosome Location:
          Chr.19: 5795690 - 5802671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01227912_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Malat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002847.2 1 2035 138
          GenBank mRNA AK008790.1 - 1 477 135
          BC004722.1 - 1337 138
          BC094333.1 - 1 1131 138
          EF177380.1 - 1 2035 138
          FJ209304.1 - 1 2035 138

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          nuclear speck organization
          regulation of synapse organization
          positive regulation of myoblast proliferation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021187 EF177380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021187 NR_002847.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74882 - -
          Whole Mouse Genome G4122A A_52_P392598 EF177380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392598 AK020134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392598 NR_002847.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394383 EF177380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394383 BC004722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394383 NR_002847.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930896 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465245 - -
          Affymetrix HT MG-430 PM Array Plate 1452378_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427285_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1429060_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438403_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1418189_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1436202_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418188_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465244 - -
          Mouse Genome 430 2.0 Array 1438403_s_at - -
          Mouse Genome 430 2.0 Array 1436202_PM_at - -
          Mouse Genome 430 2.0 Array 1427285_PM_s_at - -
          Mouse Genome 430 2.0 Array 1452378_at - -
          Mouse Genome 430 2.0 Array 1438403_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427285_s_at - -
          Mouse Genome 430 2.0 Array 1452378_PM_at - -
          Mouse Genome 430 2.0 Array 1436202_at - -
          Affymetrix Mouse Clariom S Array PSR1900008944.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008944.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008929.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008929.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008916.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008916.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008933.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008933.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008940.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008940.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008939.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008939.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008922.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008922.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008910.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008910.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008934.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008934.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008923.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008923.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008920.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008920.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008905.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008905.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008915.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008915.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008914.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008914.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008942.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008942.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008919.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008919.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008891.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008891.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008918.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008918.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008900.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008900.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008904.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008904.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008899.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008899.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008913.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008913.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008924.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008924.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008895.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008895.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008907.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008907.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008908.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008908.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008902.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008902.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008945.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008945.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008893.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008893.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008930.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008930.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008894.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008894.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008901.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008901.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008925.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008925.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008926.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008926.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008941.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008941.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008903.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008903.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008935.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008935.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008911.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008911.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008906.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008906.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008917.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008917.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008912.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008912.mm.1 TC1900000984.mm.1 MTA TC
          Mouse Expression Set 430 1452378_PM_at - -
          Mouse Expression Set 430 1438403_s_at - -
          Mouse Expression Set 430 1427285_PM_s_at - -
          Mouse Expression Set 430 1427285_s_at - -
          Mouse Expression Set 430 1438403_PM_s_at - -
          Mouse Expression Set 430 1452378_at - -
          Mouse Expression Set 430 1436202_PM_at - -
          Mouse Expression Set 430 1436202_at - -
          Mouse Genome 430A 2.0 Array 1438403_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452378_at - -
          Mouse Genome 430A 2.0 Array 1427285_s_at - -
          Mouse Genome 430A 2.0 Array 1452378_PM_at - -
          Mouse Genome 430A 2.0 Array 1427285_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1438403_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418189_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452378_at - -
          Affymetrix HT Mouse Genome 430A Array 1418188_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418189_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438403_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452378_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427285_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438403_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418188_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427285_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1429060_at - -
          Affymetrix HT Mouse Genome 430B Array 1429060_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436202_at - -
          Affymetrix HT Mouse Genome 430B Array 1436202_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 713193 - -
          ABI Mouse Genome Survey Microarray 795622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline