Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01228299_m1
          See other LYG1 GE Assays ›
          Gene Symbol:
          Lyg1
          Gene Name:
          lysozyme G-like 1
          Gene Aliases:
          2300002O18Rik
          Chromosome Location:
          Chr.1: 37946738 - 37957759 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01228299_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027111.3 NP_081387.1 4 - 5 393 61
          GenBank mRNA AK009014.1 - 4 - 5 394 61
          BC119394.1 - 4 - 5 386 8734143 61
          BC119396.1 - 4 - 5 386 8734145 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002212.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metabolic process
          peptidoglycan catabolic process
          cell wall macromolecule catabolic process
          molecular_function
          lysozyme activity
          lytic transglycosylase activity
          mannosidase activity
          mannosyl-oligosaccharide mannosidase activity
          galactosidase activity
          glucosidase activity
          trehalase activity
          fucosidase activity
          hexosaminidase activity
          amylase activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          dextrin alpha-glucosidase activity
          starch alpha-glucosidase activity
          beta-glucanase activity
          beta-6-sulfate-N-acetylglucosaminidase activity
          glucan endo-1,4-beta-glucosidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954945 NM_027111.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954945 AK009014.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116716 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119587 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954945 NM_027111.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954945 AK009014.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00047344 - -
          Whole Mouse Genome G4122A A_52_P654646 NM_027111.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654646 AK009014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654646 BC119394.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654646 BC119396.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20156731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00274463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00385402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00988409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00617694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01090579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00789884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00385400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00296870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00446153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00789885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20156800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00789881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00403024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00403023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00385403 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10354074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354075 - -
          Affymetrix HT MG-430 PM Array Plate 1429334_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10354072 - -
          Mouse Genome 430 2.0 Array 1429334_at - -
          Mouse Genome 430 2.0 Array 1429334_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69541_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69541_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100017833.mm.1 NM_027111.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017833.mm.1 NM_027111.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017833.mm.1 TC0100002212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017828.mm.1 NM_027111.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017828.mm.1 NM_027111.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017828.mm.1 TC0100002212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017830.mm.1 NM_027111.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017830.mm.1 NM_027111.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017830.mm.1 TC0100002212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017834.mm.1 NM_027111.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017834.mm.1 NM_027111.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017834.mm.1 TC0100002212.mm.1 MTA TC
          Mouse Expression Set 430 1429334_PM_at - -
          Mouse Expression Set 430 1429334_at - -
          Murine Genome U74v2 Set 165496_at - -
          Murine Genome U74v2 Set 165496_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429334_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429334_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 670089 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674398 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674398 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline