Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231041_g1
          See other UQCC3 GE Assays ›
          Gene Symbol:
          Uqcc3
          Gene Name:
          ubiquinol-cytochrome c reductase complex assembly factor 3
          Gene Aliases:
          AI462493, Cbp4
          Chromosome Location:
          Chr.19: 8880014 - 8880933 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231041_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uqcc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160356.1 NP_001153828.1 1 - 2 223 64
          GenBank mRNA AK131602.1 - 1 - 2 214 64
          AK131892.1 - 1 - 2 187 64
          AK132116.1 - 1 - 2 214 64
          AY941793.1 - 1 - 2 214 64
          BC029863.1 - 1 - 2 148 4460168 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001073.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 BC029863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 BQ174007.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 NM_001160356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 AK131602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 AK131892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 BC029863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 BQ174007.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 NM_001160356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 AK131602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453246 AK131892.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613006 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133227 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 BC029863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 BQ174007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 NM_001160356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 AK131602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 AK131892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 BC029863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 BQ174007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 NM_001160356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 AK131602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453246 AK131892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 BC029863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 BQ174007.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 NM_001160356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 AK131602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 AK131892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 BC029863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 BQ174007.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 NM_001160356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 AK131602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453246 AK131892.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465827 - -
          Affymetrix HT MG-430 PM Array Plate 1455152_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465826 - -
          Mouse Genome 430 2.0 Array 1455152_PM_at - -
          Mouse Genome 430 2.0 Array 1455152_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107197_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107197_at - -
          Affymetrix Mouse Clariom S Array PSR1900010065.mm.1 NM_001160356.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010065.mm.1 NM_001160356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010065.mm.1 TC1900001073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010063.mm.1 NM_001160356.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010063.mm.1 NM_001160356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010063.mm.1 TC1900001073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010064.mm.1 NM_001160356.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010064.mm.1 NM_001160356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010064.mm.1 TC1900001073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010066.mm.1 NM_001160356.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010066.mm.1 NM_001160356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010066.mm.1 TC1900001073.mm.1 MTA TC
          Mouse Expression Set 430 1455152_PM_at - -
          Mouse Expression Set 430 1455152_at - -
          Murine Genome U74v2 Set 94459_at - -
          Murine Genome U74v2 Set 94459_PM_at - -
          Mouse Genome 430A 2.0 Array 1455152_PM_at - -
          Mouse Genome 430A 2.0 Array 1455152_at - -
          Affymetrix HT Mouse Genome 430A Array 1455152_at - -
          Affymetrix HT Mouse Genome 430A Array 1455152_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 797634 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227303 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline