Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231452_g1
          See other DRAP1 GE Assays ›
          Gene Symbol:
          Drap1
          Gene Name:
          Dr1 associated protein 1 (negative cofactor 2 alpha)
          Gene Aliases:
          2310074H19Rik, NC2-alpha
          Chromosome Location:
          Chr.19: 5422804 - 5424979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231452_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Drap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291080.1 NP_001278009.1 6 - 7 804 77
          NM_024176.2 NP_077138.1 6 - 7 783 77
          GenBank mRNA AK005445.1 - 5 - 6 518 77
          AK010152.1 - 6 - 7 737 77
          AK087456.1 - 6 - 7 609 77
          AK154251.1 - 5 - 6 563 77
          BC002090.1 - 6 - 7 720 3488139 77
          CT010290.1 - 6 - 7 518 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000964.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Drap1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928602 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928605 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928603 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982096 BC002090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982096 NM_024176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982096 AK005445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982096 AK010152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982096 AK087456.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227765 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164602 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982096 BC002090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982096 NM_024176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982096 AK005445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982096 AK010152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982096 AK087456.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479933 BC002090.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479933 CT010290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479933 NM_024176.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479933 AK005445.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479933 AK010152.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928592 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164602 - -
          Whole Mouse Genome G4122A A_51_P479933 BC002090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479933 CT010290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479933 NM_024176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479933 AK005445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479933 AK010152.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111143 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164602 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14319018 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14262678 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928603 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928605 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14354728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14437391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14499308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14354732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14332373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14354729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465047 - -
          Affymetrix HT MG-430 PM Array Plate 1451676_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465045 - -
          Mouse Genome 430 2.0 Array 1451676_PM_at - -
          Mouse Genome 430 2.0 Array 1451676_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66556_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66556_at - -
          Affymetrix Mouse Clariom S Array PSR1900008568.mm.1 NM_024176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008568.mm.1 TC1900000964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008569.mm.1 NM_024176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008569.mm.1 TC1900000964.mm.1 MTA TC
          Mouse Expression Set 430 1451676_at - -
          Mouse Expression Set 430 1451676_PM_at - -
          Murine Genome U74v2 Set 96617_at - -
          Murine Genome U74v2 Set 96617_PM_at - -
          Mouse Genome 430A 2.0 Array 1451676_at - -
          Mouse Genome 430A 2.0 Array 1451676_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907754 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2940142 - -
          Illumina mouse-6 v1.0 expression beadchip 2940142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639703 - -
          mouse-6 v1.1 (Illumina) scl53495.6.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53495.6.1_70-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639703 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53495.6.1_70-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline