Search
Search

Snx32| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024560.2 | NP_001019731.1 | 1 - 2 | 156 | 85 | |
| GenBank mRNA | AK037118.1 | - | 163 | 85 | ||
| AK047234.1 | - | 1 - 2 | 131 | 85 | ||
| AK143441.1 | - | 1 - 2 | 174 | 85 | ||
| BC048922.1 | - | 1 - 2 | 156 | 6311864 | 85 | |
Set Membership: |
|