Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231512_m1
          See other KAT5 GE Assays ›
          Gene Symbol:
          Kat5
          Gene Name:
          K(lysine) acetyltransferase 5
          Gene Aliases:
          AI839539, CPLA2, Htatip, Htatip1, PLIP, Tip55, Tip60
          Chromosome Location:
          Chr.19: 5603014 - 5610232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231512_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kat5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199247.1 NP_001186176.1 8 - 9 842 86
          NM_001199248.1 NP_001186177.1 7 - 8 941 86
          NM_001199249.1 NP_001186178.1 7 - 8 941 86
          NM_178637.2 NP_848752.1 9 - 10 998 86
          NR_037603.1 6 - 7 874 86
          GenBank mRNA AB055409.1 - 6 - 7 852 86
          AF528194.1 - 5 - 6 552 86
          AF528195.1 - 8 - 9 933 86
          AF528196.1 - 7 - 8 777 86
          AK143674.1 - 7 - 8 893 86
          AK146352.1 - 9 - 10 955 86
          AK159568.1 - 7 - 8 874 86
          AK160077.1 - 7 - 8 874 86
          AK219598.1 - 2 - 3 219 86
          BC110675.1 - 9 - 10 939 4011689 86
          BC129967.1 - 7 - 8 776 86
          BC129968.1 - 7 - 8 777 40106268 86
          CA317697.1 - 2 - 3 236 86
          DQ139980.1 - 8 - 9 933 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000975.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kat5

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          double-strand break repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          response to ionizing radiation
          chromatin modification
          histone acetylation
          negative regulation of interleukin-2 production
          regulation of growth
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to estradiol stimulus
          positive regulation of protein acetylation
          chromatin binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          transcription factor binding
          transferase activity
          transferase activity, transferring acyl groups
          transferase activity, transferring acyl groups other than amino-acyl groups
          protein complex binding
          phospholipase binding
          metal ion binding
          repressing transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199247.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199247.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199248.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199248.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199249.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_001199249.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_178637.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NM_178637.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NR_037603.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 NR_037603.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008699.mm.1 TC1900000975.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0