Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231591_m1
          See other BATF2 GE Assays ›
          Gene Symbol:
          Batf2
          Gene Name:
          basic leucine zipper transcription factor, ATF-like 2
          Gene Aliases:
          4933430F08Rik
          Chromosome Location:
          Chr.19: 6164392 - 6172476 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231591_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Batf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028967.1 NP_083243.1 2 - 3 202 72
          GenBank mRNA AK016990.1 - 418 72
          AK150133.1 - 2 - 3 222 72
          BC024521.1 - 2 - 3 202 4984403 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000102.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Batf2

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell differentiation
          defense response to protozoan
          myeloid dendritic cell differentiation
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142614 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160774 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165182 AK016990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09095567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03544255 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03544252 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09095590 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09095529 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09095541 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160774 - -
          Whole Mouse Genome G4122A A_51_P165182 AK016990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165182 AK016990.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101396 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09095541 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14467162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14163889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14373419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14254901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14318917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14467164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14499771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14440849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14190338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14499772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14278633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14476892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14476897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14467172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14467169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14472349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14420200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14318926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14254895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933059 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460769 - -
          Affymetrix HT MG-430 PM Array Plate 1430005_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460767 - -
          Mouse Genome 430 2.0 Array 1430005_PM_a_at - -
          Mouse Genome 430 2.0 Array 1430005_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74481_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74481_at - -
          Affymetrix Mouse Clariom S Array PSR1900001189.mm.1 NM_028967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001189.mm.1 NM_028967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001189.mm.1 TC1900000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001190.mm.1 NM_028967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001190.mm.1 NM_028967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001190.mm.1 TC1900000102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001192.mm.1 NM_028967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001192.mm.1 NM_028967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001192.mm.1 TC1900000102.mm.1 MTA TC
          Mouse Expression Set 430 1430005_PM_a_at - -
          Mouse Expression Set 430 1430005_a_at - -
          Mouse Genome 430A 2.0 Array 1430005_a_at - -
          Mouse Genome 430A 2.0 Array 1430005_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430005_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430005_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 649558 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980100 - -
          Illumina mouse-6 v1.0 expression beadchip 1980100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230143 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230143 - -
          mouse-6 v1.1 (Illumina) scl52865.6_503-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52865.6_503-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230143 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline