| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900024819.mm.1 |
NM_001167923.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024819.mm.1 |
NM_001167923.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024819.mm.1 |
TC0900002999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024815.mm.1 |
NM_001167923.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024815.mm.1 |
NM_001167923.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024815.mm.1 |
TC0900002999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024816.mm.1 |
NM_001167923.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024816.mm.1 |
NM_001167923.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024816.mm.1 |
TC0900002999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024817.mm.1 |
NM_001167923.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024817.mm.1 |
NM_001167923.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024817.mm.1 |
TC0900002999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024818.mm.1 |
NM_001167923.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024818.mm.1 |
NM_001167923.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024818.mm.1 |
TC0900002999.mm.1 |
MTA TC |