Search
Search

Yipf4| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026417.4 | NP_080693.2 | 3 - 4 | 633 | 124 | |
| GenBank mRNA | AK017721.1 | - | 3 - 4 | 611 | 124 | |
| AK017722.1 | - | 3 - 4 | 611 | 124 | ||
| AK083301.1 | - | 3 - 4 | 613 | 124 | ||
| AK159227.1 | - | 3 - 4 | 594 | 124 | ||
| AK166347.1 | - | 3 - 4 | 621 | 124 | ||
| BC003936.1 | - | 4 - 5 | 808 | 3486503 | 124 | |
Set Membership: |
|