Search
Search

Klhdc8b| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030075.2 | NP_084351.1 | 7 - 8 | 1612 | 69 | |
| GenBank mRNA | AK019849.1 | - | 7 - 8 | 1610 | 69 | |
| AK154428.1 | - | 7 - 8 | 1713 | 69 | ||
| BC048918.1 | - | 2 - 3 | 575 | 6309434 | 69 | |
Set Membership: |
|