| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700023419.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023419.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023419.mm.1 |
TC1700002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023423.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023423.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023423.mm.1 |
TC1700002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023425.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023425.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023425.mm.1 |
TC1700002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023422.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023422.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023422.mm.1 |
TC1700002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023426.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023426.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023426.mm.1 |
TC1700002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700023420.mm.1 |
NM_001163625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023420.mm.1 |
NM_001163625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700023420.mm.1 |
TC1700002559.mm.1 |
MTA TC |