Search
Search

Srsf7| Gene Symbol: Srsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195485.1 | NP_001182414.1 | 3 - 4 | 485 | 64 | |
| NM_001195486.1 | NP_001182415.1 | 3 - 4 | 485 | 64 | ||
| NM_001195487.1 | NP_001182416.1 | 3 - 4 | 485 | 64 | ||
| NM_146083.2 | NP_666195.1 | 3 - 4 | 485 | 64 | ||
| GenBank mRNA | AI550251.1 | - | 1 - 2 | 63 | 64 | |
| AK045884.1 | - | 3 - 4 | 647 | 64 | ||
| AK082848.1 | - | 3 - 4 | 485 | 64 | ||
| AK137795.1 | - | 3 - 4 | 486 | 64 | ||
| AK161652.1 | - | 3 - 4 | 484 | 64 | ||
| AK168355.1 | - | 3 - 4 | 485 | 64 | ||
| BC014857.1 | - | 3 - 4 | 466 | 2646366 | 64 | |
| BC025529.1 | - | 3 - 4 | 476 | 5342587 | 64 | |
| BY740981.1 | - | 3 - 4 | 484 | 64 | ||
Set Membership: |
|